Diphenyl-N-cyanocarbonimidate 97%
Dodavatel: Apollo Scientific
Synonyms:
Diphenyl N-cyanocarbonimidate, N-Cyanocarbonimidic Acid Diphenyl Ester
OR16450-25G
APOSOR16450-25GEA
3190
CZK
APOSOR16450-25G
Diphenyl-N-cyanocarbonimidate 97%
Diphenyl-N-cyanocarbonimidate
Vzorec:
C₁₄H₁₀N₂O₂ MW: 238,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010380 CAS číslo: 79463-77-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...