Tisk…

6-Amino-2-methylnicotinic acid

Dodavatel: Apollo Scientific
Varování

OR16448-1G OR16448-100MG OR16448-250MG OR16448-500MG OR16448-2.5G
APOSOR16448-1GEA 12300 CZK
APOSOR16448-1G APOSOR16448-100MG APOSOR16448-250MG APOSOR16448-500MG APOSOR16448-2.5G
6-Amino-2-methylnicotinic acid
6-Amino-2-methylnicotinic acid
Vzorec: C₇H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09258839
CAS číslo: 680208-82-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR