Diphenyl phosphate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Diphenyl hydrogen phosphate
OR16436-100G
OR16436-25G
OR16436-5G
APOSOR16436-100GEA
2360
CZK
APOSOR16436-100G
APOSOR16436-25G
APOSOR16436-5G
Diphenyl phosphate
Diphenyl phosphate
Vzorec:
C₁₂H₁₁O₄P MW: 250,19 g/mol Bod tání: 62…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003033 CAS číslo: 838-85-7 EINECS: 212-357-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...