Tisk…

2-(3,4-Dichlorophenyl)-2-oxoacetaldehyde hydrate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,4-Dichlorophenylglyoxal hydrate

OR1627-1G OR1627-25G OR1627-5G
APOSOR1627-1GEA 2900 CZK
APOSOR1627-1G APOSOR1627-25G APOSOR1627-5G
2-(3,4-Dichlorophenyl)-2-oxoacetaldehyde hydrate 95%
2-(3,4-Dichlorophenyl)-2-oxoacetaldehyde hydrate
Vzorec: C₈H₆Cl₂O₃
Storage Temperature: Ambient
CAS číslo: 859775-23-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR