Tisk…

2-(Methylsulphinyl)benzenecarboxylic acid

Dodavatel: Apollo Scientific

Synonyms: 2-(Methylsulphinyl)benzoic acid

OR16149-1G OR16149-250MG OR16149-500MG
APOSOR16149-1GEA 2610 CZK
APOSOR16149-1G APOSOR16149-250MG APOSOR16149-500MG
2-(Methylsulphinyl)benzenecarboxylic acid
2-(Methylsulphinyl)benzenecarboxylic acid
Vzorec: C₈H₈O₃S
Bod tání: 150…178 °C
Storage Temperature: Ambient
MDL Number: MFCD01936003
CAS číslo: 19093-34-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR