2-Ethylbenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR16146-100G
OR16146-25G
OR16146-5G
APOSOR16146-100GEA
4240
CZK
APOSOR16146-100G
APOSOR16146-25G
APOSOR16146-5G
2-Ethylbenzoic acid 98%
2-Ethylbenzoic acid
Vzorec:
C₉H₁₀O₂ Bod tání: 62…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045838 CAS číslo: 612-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...