Oxo[4-(piperidin-1-yl)phenyl]acetaldehyde hydrate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-[4-(2-Oxoethanoyl)phenyl]piperidine hydrate, 4-(Piperidin-1-yl)phenylglyoxal hydrate
OR1613-1G
OR1613-500MG
OR1613-5G
APOSOR1613-1GEA
4960
CZK
APOSOR1613-1G
APOSOR1613-500MG
APOSOR1613-5G
Oxo[4-(piperidin-1-yl)phenyl]acetaldehyde hydrate 95%
Oxo[4-(piperidin-1-yl)phenyl]acetaldehyde hydrate
Vzorec:
C₁₃H₁₅NO₂.H₂O Storage Temperature: Ambient |
MDL Number:
MFCD08705865 CAS číslo: 93290-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...