Tisk…

2-Nitro-4-picoline

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Methyl-2-nitropyridine

OR16077-1G OR16077-5G
APOSOR16077-1GEA 2110 CZK
APOSOR16077-1G APOSOR16077-5G
2-Nitro-4-picoline
2-Nitro-4-picoline
Vzorec: C₆H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04114155
CAS číslo: 18368-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR