Tisk…

1-Acetyl-4-(methylamino)piperidine

Dodavatel: Apollo Scientific
Nebezpečí

OR16036-1G OR16036-250MG OR16036-500MG OR16036-2.5G
APOSOR16036-1GEA 1620 CZK
APOSOR16036-1G APOSOR16036-250MG APOSOR16036-500MG APOSOR16036-2.5G
1-Acetyl-4-(methylamino)piperidine
1-Acetyl-4-(methylamino)piperidine
Vzorec: C₈H₁₆N₂O
MW: 156,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367773
CAS číslo: 139062-96-7
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR