Tisk…

4-Methoxyphenylglyoxal hydrate 95%

Dodavatel: Apollo Scientific

OR1600-1G OR1600-5G
APOSOR1600-1GEA 0 CZK
APOSOR1600-1G APOSOR1600-5G
4-Methoxyphenylglyoxal hydrate 95%
4-Methoxyphenylglyoxal hydrate
Vzorec: C₉H₁₀O₄
Bod varu: 145…147 °C/13mmHg
Bod tání: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00234534
CAS číslo: 16208-17-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR