4-Bromo-3-chlorotoluene
Dodavatel: Apollo Scientific
|
Varování
|
OR1598-25G
OR1598-5G
APOSOR1598-25GEA
705
CZK
APOSOR1598-25G
APOSOR1598-5G
4-Bromo-3-chlorotoluene
4-Bromo-3-chlorotoluene
Vzorec:
C₇H₆BrCl MW: 205,48 g/mol Bod varu: 100 °C/22mmHg Hustota: 1,54 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00045161 CAS číslo: 6627-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...