Tisk…

4-Hydroxyphenylglyoxal hydrate 95%

Dodavatel: Apollo Scientific
Varování

OR1597-1G OR1597-5G
APOSOR1597-1GEA 6220 CZK
APOSOR1597-1G APOSOR1597-5G
4-Hydroxyphenylglyoxal hydrate 95%
4-Hydroxyphenylglyoxal hydrate
Vzorec: C₈H₈O₄
Storage Temperature: Ambient
CAS číslo: 197447-05-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR