Tisk…

2,4-Dichloro-5-nitropyridine 97%

Dodavatel: Apollo Scientific
Varování

OR15939-1G OR15939-5G OR15939-25G
APOSOR15939-1GEA 787 CZK
APOSOR15939-1G APOSOR15939-5G APOSOR15939-25G
2,4-Dichloro-5-nitropyridine 97%
2,4-Dichloro-5-nitropyridine
Vzorec: C₅H₂Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07368834
CAS číslo: 4487-56-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR