Tisk…

3-(4'-Chlorophenyl)propyl chloride

Dodavatel: Apollo Scientific

OR1591-1G OR1591-5G OR1591-10G
APOSOR1591-1GEA 2110 CZK
APOSOR1591-1G APOSOR1591-5G APOSOR1591-10G
3-(4'-Chlorophenyl)propyl chloride
3-(4'-Chlorophenyl)propyl chloride
Vzorec: C₉H₁₀Cl₂
Bod varu: 135…138 °C/0,15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00045302
CAS číslo: 64473-34-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR