Tisk…

7-Bromobenzo[b]furan 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 7-Bromobenzofuran

OR15756-1G OR15756-250MG
APOSOR15756-1GEA 705 CZK
APOSOR15756-1G APOSOR15756-250MG
7-Bromobenzo[b]furan 95%
7-Bromobenzo[b]furan
Vzorec: C₈H₅BrO
MW: 197,031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09056756
CAS číslo: 133720-60-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR