Tisk…

Iron(III) acetylacetonate

Dodavatel: Apollo Scientific
Varování

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

OR15745-100G OR15745-500G
APOSOR15745-100GEA 1870 CZK
APOSOR15745-100G APOSOR15745-500G
Iron(III) acetylacetonate
Iron(III) acetylacetonate
Vzorec: C₁₅H₂₁FeO₆
MW: 353.18
Bod varu: 100
Bod tání: 180…182 °C
Hustota: 5.24 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000020
CAS číslo: 14024-18-1
EINECS: 237-853-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR