Tisk…

(S)-(+)-1-Boc-3-hydroxypyrrolidine 99%

Dodavatel: Apollo Scientific
Varování

OR15680-100G OR15680-10G OR15680-25G
APOSOR15680-100GEA 2440 CZK
APOSOR15680-100G APOSOR15680-10G APOSOR15680-25G
(S)-(+)-1-Boc-3-hydroxypyrrolidine 99%
(S)-(+)-1-Boc-3-hydroxypyrrolidine

Chiral intermediate for pharmaceuticals and agrochemicals.

Vzorec: C₉H₁₇NO₃
Bod tání: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01317839
CAS číslo: 101469-92-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR