Tisk…

3-Formyl-N-(tert-butoxycarbonyl)piperidine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Piperidine-3-carboxaldehyde, N-BOC protected, tert-Butyl 3-formylpiperidine-1-carboxylate

OR15677-5G OR15677-25G OR15677-100G
APOSOR15677-5GEA 1120 CZK
APOSOR15677-5G APOSOR15677-25G APOSOR15677-100G
3-Formyl-N-(tert-butoxycarbonyl)piperidine 98%
3-Formyl-N-(tert-butoxycarbonyl)piperidine
Vzorec: C₁₁H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD02179020
CAS číslo: 118156-93-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR