1-Boc-4-(methylamino)piperidine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-(Methylamino)piperidine, N1-BOC protected, tert-Butyl 4-(methylamino)piperidine-1-carboxylate
OR15676-25G
OR15676-100G
APOSOR15676-25GEA
1870
CZK
APOSOR15676-25G
APOSOR15676-100G
1-Boc-4-(methylamino)piperidine
1-Boc-4-(methylamino)piperidine
Vzorec:
C₁₁H₂₂N₂O₂ MW: 214,3046 g/mol Hustota: 1,005 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02259411 CAS číslo: 147539-41-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...