(S)-N-Boc-3-piperidinemethanol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(s)-n-(tert-butoxycarbonyl)-3-hydroxymethylpiperidine, (s)-tert-butyl 3-(hydroxymethyl)tetrahydro-1(2h)-pyridinecarboxylate, (S)-1-BOC-3-(Hydroxymethyl)piperidine, tert-Butyl (S)-3-(hydroxymethyl)piperidine-1-carboxylate
OR15674-1G
OR15674-25G
OR15674-5G
APOSOR15674-1GEA
622
CZK
APOSOR15674-1G
APOSOR15674-25G
APOSOR15674-5G
(S)-N-Boc-3-piperidinemethanol
(S)-N-Boc-3-piperidinemethanol
Vzorec:
C₁₁H₂₁NO₃ MW: 215,293 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02683203 CAS číslo: 140695-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...