(S)-3-(Aminomethyl)-1-boc-piperidine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl (S)-3-(aminomethyl)piperidine-1-carboxylate, (S)-1-BOC-3-(Aminomethyl)piperidine
OR15672-1G
OR15672-5G
APOSOR15672-1GEA
1040
CZK
APOSOR15672-1G
APOSOR15672-5G
(S)-3-(Aminomethyl)-1-boc-piperidine 97%
(S)-3-(Aminomethyl)-1-boc-piperidine
Vzorec:
C₁₁H₂₂N₂O₂ MW: 214,31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03839877 CAS číslo: 140645-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...