Tisk…

1-Iodo-4-methylimidazolium chloride

Dodavatel: Apollo Scientific

Synonyms: 4-Iodo-1-methyl-1H-imidazole hydrochloride

OR15576-1G OR15576-5G
APOSOR15576-1GEA 8700 CZK
APOSOR15576-1G APOSOR15576-5G
1-Iodo-4-methylimidazolium chloride
1-Iodo-4-methylimidazolium chloride
Vzorec: C₄H₆ClIN₂
Storage Temperature: Ambient
MDL Number: MFCD07783893
CAS číslo: 871333-96-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR