4-Iodo-2-(methylthio)pyrimidine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Iodo-2-methylsulphanylpyrimidine
OR15525-1G
OR15525-5G
APOSOR15525-1GEA
2690
CZK
APOSOR15525-1G
APOSOR15525-5G
4-Iodo-2-(methylthio)pyrimidine
4-Iodo-2-(methylthio)pyrimidine
Vzorec:
C₅H₅IN₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD01319020 CAS číslo: 1122-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...