2-(1-Phenylethylidene)-1-hydrazinecarboxamide
Dodavatel: Apollo Scientific
OR15508-10G
OR15508-1G
OR15508-5G
APOSOR15508-10GEA
14190
CZK
APOSOR15508-10G
APOSOR15508-1G
APOSOR15508-5G
2-(1-Phenylethylidene)-1-hydrazinecarboxamide
2-(1-Phenylethylidene)-1-hydrazinecarboxamide
Vzorec:
C₉H₁₁N₃O MW: 177,21 g/mol Bod tání: 207…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD00137159 CAS číslo: 2492-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...