Methyl-4-bromo-1H-pyrrole-2-carboxylate 97+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 4-bromopyrrole-2-carboxylate, Methyl 4-bromo-1H-pyrrole-2-carboxylate
OR15416-1G
OR15416-5G
APOSOR15416-1GEA
622
CZK
APOSOR15416-1G
APOSOR15416-5G
Methyl-4-bromo-1H-pyrrole-2-carboxylate 97+%
Methyl-4-bromo-1H-pyrrole-2-carboxylate
Vzorec:
C₆H₆BrNO₂ MW: 204,00 g/mol Bod tání: 95…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00832855 CAS číslo: 934-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...