2-(4-Hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid 95%
Dodavatel: Apollo Scientific
OR15412-10G
OR15412-1G
OR15412-5G
APOSOR15412-10GEA
25460
CZK
APOSOR15412-10G
APOSOR15412-1G
APOSOR15412-5G
2-(4-Hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid 95%
2-(4-Hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid
Vzorec:
C₁₇H₁₃NO₃ MW: 279,3 g/mol Bod tání: 338 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200949 CAS číslo: 107419-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...