Tisk…

2-Acetyl-3-chlorothiophene

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(3-Chloro-2-thienyl)-1-ethanone

OR15400-1G OR15400-5G
APOSOR15400-1GEA 1330 CZK
APOSOR15400-1G APOSOR15400-5G
2-Acetyl-3-chlorothiophene
2-Acetyl-3-chlorothiophene
Vzorec: C₆H₅ClOS
MW: 160,62 g/mol
Bod varu: 117…118 °C/15mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00794248
CAS číslo: 89581-82-8
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR