6-Chloro-2-(3-chlorophenyl)-3-methyl-4-quinolinecarboxylic acid
Dodavatel: Apollo Scientific
OR15390-10G
OR15390-1G
OR15390-5G
APOSOR15390-10GEA
25460
CZK
APOSOR15390-10G
APOSOR15390-1G
APOSOR15390-5G
6-Chloro-2-(3-chlorophenyl)-3-methyl-4-quinolinecarboxylic acid
6-Chloro-2-(3-chlorophenyl)-3-methyl-4-quinolinecarboxylic acid
Vzorec:
C₁₇H₁₁Cl₂NO₂ MW: 332,19 g/mol Bod tání: 324 °C Storage Temperature: Ambient |
MDL Number:
MFCD03420193 CAS číslo: 886361-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...