2-Amino-5-(4-chlorophenyl)-4-thiazolecarbohydrazide
Dodavatel: Apollo Scientific
OR15386-10G
OR15386-1G
OR15386-5G
APOSOR15386-10GEA
38350
CZK
APOSOR15386-10G
APOSOR15386-1G
APOSOR15386-5G
2-Amino-5-(4-chlorophenyl)-4-thiazolecarbohydrazide
2-Amino-5-(4-chlorophenyl)-4-thiazolecarbohydrazide
Vzorec:
C₁₀H₉ClN₄OS Bod tání: 257 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200937 CAS číslo: 886361-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...