Tisk…

1-(4-Bromo-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloro-1-ethanone

Dodavatel: Apollo Scientific

Synonyms: 4-Bromo-1-methyl-2-(trichloroacetyl)-1H-pyrrole

OR15312-10G OR15312-1G OR15312-5G
APOSOR15312-10GEA 14060 CZK
APOSOR15312-10G APOSOR15312-1G APOSOR15312-5G
1-(4-Bromo-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloro-1-ethanone
1-(4-Bromo-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloro-1-ethanone
Vzorec: C₇H₅BrCl₃NO
MW: 305,39 g/mol
Bod tání: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD08276938
CAS číslo: 184643-69-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR