Tisk…

4-(4-Ethylpiperazin-1-yl)phenylamine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(4-Ethylpiperazin-1-yl)aniline

OR15287-1G OR15287-5G
APOSOR15287-1GEA 622 CZK
APOSOR15287-1G APOSOR15287-5G
4-(4-Ethylpiperazin-1-yl)phenylamine 98%
4-(4-Ethylpiperazin-1-yl)phenylamine
Vzorec: C₁₂H₁₉N₃
MW: 205,3000 g/mol
Bod tání: 76…78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00702246
CAS číslo: 115619-01-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR