2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR15233-1G
OR15233-5G
APOSOR15233-1GEA
6620
CZK
APOSOR15233-1G
APOSOR15233-5G
2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid
2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid
Vzorec:
C₁₂H₁₁NO₂S Bod tání: 135…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00277218 CAS číslo: 101736-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...