Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate
Dodavatel: Apollo Scientific
OR15185-10G
OR15185-1G
OR15185-5G
APOSOR15185-10GEA
10910
CZK
APOSOR15185-10G
APOSOR15185-1G
APOSOR15185-5G
Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate
Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate
Vzorec:
C₁₆H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD01764136 CAS číslo: 279692-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...