Tisk…

2,6-Bis[(dimethylamino)methyl]-4-(2-methylbutyl)benzenol

Dodavatel: Apollo Scientific

Synonyms: 2,6-Bis[(dimethylamino)methyl]-4-(2-methylbutyl)phenol

OR15181-1G OR15181-5G
APOSOR15181-1GEA 7670 CZK
APOSOR15181-1G APOSOR15181-5G
2,6-Bis[(dimethylamino)methyl]-4-(2-methylbutyl)benzenol
2,6-Bis[(dimethylamino)methyl]-4-(2-methylbutyl)benzenol
Vzorec: C₁₇H₃₀N₂O
MW: 278,44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443977
CAS číslo: 924868-92-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR