Tisk…

2,6-Bis[(dimethylamino)methyl]-4-ethylbenzenol

Dodavatel: Apollo Scientific

Synonyms: 2,6-Bis[(dimethylamino)methyl]-4-ethylphenol

OR15179-10G OR15179-1G OR15179-5G
APOSOR15179-10GEA 38350 CZK
APOSOR15179-10G APOSOR15179-1G APOSOR15179-5G
2,6-Bis[(dimethylamino)methyl]-4-ethylbenzenol
2,6-Bis[(dimethylamino)methyl]-4-ethylbenzenol
Vzorec: C₁₄H₂₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD08443957
CAS číslo: 36805-45-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR