2-(tert-Butyl)-6-[(dimethylamino)methyl]-4-methylbenzenol
Dodavatel: Apollo Scientific
Synonyms:
2-(tert-Butyl)-6-[(dimethylamino)methyl]-4-methylphenol
OR15145-10G
OR15145-1G
OR15145-5G
APOSOR15145-10GEA
25460
CZK
APOSOR15145-10G
APOSOR15145-1G
APOSOR15145-5G
2-(tert-Butyl)-6-[(dimethylamino)methyl]-4-methylbenzenol
2-(tert-Butyl)-6-[(dimethylamino)methyl]-4-methylbenzenol
Vzorec:
C₁₄H₂₃NO MW: 221,35 g/mol Bod tání: 46…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD08443981 CAS číslo: 4142-59-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...