Tisk…

4-Bromo-3-(tert-butyl)-1H-pyrazol-5-ol

Dodavatel: Apollo Scientific

Synonyms: 4-Bromo-3-(tert-butyl)-5-hydroxy-1H-pyrazole

OR15144-1G OR15144-5G
APOSOR15144-1GEA 15310 CZK
APOSOR15144-1G APOSOR15144-5G
4-Bromo-3-(tert-butyl)-1H-pyrazol-5-ol
4-Bromo-3-(tert-butyl)-1H-pyrazol-5-ol
Vzorec: C₇H₁₁BrN₂O
MW: 219,09 g/mol
Bod tání: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD08443992
CAS číslo: 873837-19-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR