Tisk…

Methyl-2-(4-chloro-2-methylphenoxy)-3-methoxyacrylate

Dodavatel: Apollo Scientific

OR15105-1G OR15105-5G
APOSOR15105-1GEA 15310 CZK
APOSOR15105-1G APOSOR15105-5G
Methyl-2-(4-chloro-2-methylphenoxy)-3-methoxyacrylate
Methyl-2-(4-chloro-2-methylphenoxy)-3-methoxyacrylate
Vzorec: C₁₂H₁₃ClO₄
MW: 256,69 g/mol
Bod tání: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD08443943
CAS číslo: 255908-42-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR