Tisk…

{1-[(4-Bromophenyl)sulphonyl]-3-piperidinyl}methanol

Dodavatel: Apollo Scientific

OR15071-10G OR15071-1G OR15071-5G
APOSOR15071-10GEA 25460 CZK
APOSOR15071-10G APOSOR15071-1G APOSOR15071-5G
{1-[(4-Bromophenyl)sulphonyl]-3-piperidinyl}methanol
{1-[(4-Bromophenyl)sulphonyl]-3-piperidinyl}methanol
Vzorec: C₁₂H₁₆BrNO₃S
Bod tání: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD08444025
CAS číslo: 924869-08-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR