Fluoral-P (=4-amino-3-penten-2-one) 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Amino-3-penten-2-one, 4-Aminopent-3-en-2-one
OR1506-100G
APOSOR1506-100GEA
5030
CZK
APOSOR1506-100G
Fluoral-P (=4-amino-3-penten-2-one) 95%
Fluoral-P (=4-amino-3-penten-2-one)
Vzorec:
C₅H₉NO MW: 99,13 g/mol Bod varu: 130…131 °C Bod tání: 38 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043715 CAS číslo: 1118-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...