3-Chloro-5-iodobenzonitrile 99%
Dodavatel: Apollo Scientific
|
Varování
|
OR15035-1G
OR15035-25G
OR15035-5G
APOSOR15035-1GEA
622
CZK
APOSOR15035-1G
APOSOR15035-25G
APOSOR15035-5G
3-Chloro-5-iodobenzonitrile 99%
3-Chloro-5-iodobenzonitrile
Vzorec:
C₇H₃ClIN MW: 263,4629 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00672974 CAS číslo: 289039-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...