2-Chloro-5-iodobenzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR15034-25G
OR15034-5G
OR15034-100G
APOSOR15034-25GEA
2150
CZK
APOSOR15034-25G
APOSOR15034-5G
APOSOR15034-100G
2-Chloro-5-iodobenzonitrile 98%
2-Chloro-5-iodobenzonitrile
Vzorec:
C₇H₃ClIN MW: 263,4629 g/mol Bod tání: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672973 CAS číslo: 289039-29-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...