Pyridinium tribromide 90+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide
OR15023-100G
OR15023-1KG
OR15023-500G
APOSOR15023-100GEA
622
CZK
APOSOR15023-100G
APOSOR15023-1KG
APOSOR15023-500G
Pyridinium tribromide 90+%
Pyridinium tribromide
Vzorec:
C₅H₆Br₃N MW: 319,82 g/mol Bod tání: 127…133 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00013223 CAS číslo: 39416-48-3 EINECS: 254-446-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...