Tisk…

4-Bromosalicylic acid 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Bromo-2-hydroxybenzoic acid

OR15002-25G OR15002-100G OR15002-5G
APOSOR15002-25GEA 1210 CZK
APOSOR15002-25G APOSOR15002-100G APOSOR15002-5G
4-Bromosalicylic acid 98%
4-Bromosalicylic acid
Vzorec: C₇H₅BrO₃
MW: 217,02 g/mol
Bod tání: 216…217 °C
Storage Temperature: Ambient
MDL Number: MFCD06797970
CAS číslo: 1666-28-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR