6-Bromoisoquinoline
Dodavatel: Apollo Scientific
OR14998-1G
OR14998-25G
OR14998-5G
APOSOR14998-1GEA
622
CZK
APOSOR14998-1G
APOSOR14998-25G
APOSOR14998-5G
6-Bromoisoquinoline
6-Bromoisoquinoline
Vzorec:
C₉H₆BrN |
MDL Number:
MFCD04973299 CAS číslo: 34784-05-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...