Ethyl-2-chloro-2-(hydroxyimino)acetate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-2-(hydroxyimino)acetic acid ethyl ester, alpha-Chloroglyoxalic acid ethyl ester Oxime, Ethylchloroximidoacetate, Ethyl 2-Chloro-2-(hydroxyimino)acetate
OR14997-25G
OR14997-5G
OR14997-1G
APOSOR14997-25GEA
2280
CZK
APOSOR14997-25G
APOSOR14997-5G
APOSOR14997-1G
Ethyl-2-chloro-2-(hydroxyimino)acetate 97%
Ethyl-2-chloro-2-(hydroxyimino)acetate
Vzorec:
C₄H₆ClNO₃ MW: 151,5483 g/mol Bod tání: 70…76 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010209 CAS číslo: 14337-43-0 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...