Tisk…

ethylbenzen

Dodavatel: Apollo Scientific

Synonyms: Phenylethane

OR14988-100G OR14988-25G
APOSOR14988-100GEA 3110 CZK
APOSOR14988-100G APOSOR14988-25G
ethylbenzen
ethylbenzen
Vzorec: C₈H₁₀
MW: 106,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS číslo: 100-41-4
UN: 1175
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR