Tisk…

2-Amino-4-nitrobenzonitrile 95%

Dodavatel: Apollo Scientific
Varování

OR1493-1G OR1493-100MG
APOSOR1493-1GEA 705 CZK
APOSOR1493-1G APOSOR1493-100MG
2-Amino-4-nitrobenzonitrile 95%
2-Amino-4-nitrobenzonitrile
Vzorec: C₇H₅N₃O₂
MW: 163,1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04117876
CAS číslo: 87376-25-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR