Tisk…

3-Bromo-5-(chloromethyl)pyridine hydrochloride 95+%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Pyridine, 3-bromo-5-(chloromethyl)-, hydrochloride

OR14923-1G
APOSOR14923-1GEA 2150 CZK
APOSOR14923-1G
3-Bromo-5-(chloromethyl)pyridine hydrochloride 95+%
3-Bromo-5-(chloromethyl)pyridine hydrochloride
Vzorec: C₆H₆BrCl₂N
MW: 242,925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09260933
CAS číslo: 39741-46-3
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR