Tisk…

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Fmoc-D-alanine, Fmoc-D-Ala-OH

OR1492-100G OR1492-10G OR1492-25G
APOSOR1492-100GEA 1950 CZK
APOSOR1492-100G APOSOR1492-10G APOSOR1492-25G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
Vzorec: C₁₈H₁₇NO₄
MW: 311.34 g/mol
Bod tání: 151…157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062960
CAS číslo: 79990-15-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR